2-Methoxybenzoylacetonitrile
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-(2-Methoxyphenyl)-3-oxopropanenitrile, 3-(2-Methoxyphenyl)-3-oxo-propionitrile
OR7018-1G
OR7018-25G
OR7018-5G
APOSOR7018-1GEA
69.8
EUR
APOSOR7018-1G
APOSOR7018-25G
APOSOR7018-5G
2-Methoxybenzoylacetonitrile
2-Methoxybenzoylacetonitrile
Formula:
C₁₀H₉NO₂ MW: 175,184 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01993554 CAS Number: 35276-83-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...