Isopropyl laurate 98%

Supplier: Apollo Scientific
Warning

OR70188-100G OR70188-25G OR70188-5G
APOSOR70188-100GEA 33.5 EUR
APOSOR70188-100G APOSOR70188-25G APOSOR70188-5G
Isopropyl laurate 98%
Isopropyl laurate
Formula: C₁₅H₃₀O₂
Density: 0,885 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00451146
CAS Number: 10233-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR