2-Amino-4-methoxybenzenethiol

Supplier: Apollo Scientific
Warning

OR70225-1G OR70225-250MG
APOSOR70225-1GEA 780 EUR
APOSOR70225-1G APOSOR70225-250MG
2-Amino-4-methoxybenzenethiol
2-Amino-4-methoxybenzenethiol
Formula: C₇H₉NOS
Boiling Pt: 122 °C (0,5 mm Hg)
Melting Pt: 47 °C
Storage Temperature: Refrigerator
MDL Number: MFCD17014600
CAS Number: 14482-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR