4-(Propionyl)benzeneboronic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 4-(Propionyl)phenyboronic acid, (4-Propionylphenyl)boronic acid

OR7254-1G OR7254-5G OR7254-250MG
APOSOR7254-1GEA 0 EUR
APOSOR7254-1G APOSOR7254-5G APOSOR7254-250MG
4-(Propionyl)benzeneboronic acid 98%
4-(Propionyl)benzeneboronic acid
Formula: C₉H₁₁BO₃
MW: 177,993 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04039038
CAS Number: 186498-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR