3-(2-Bromophenyl)prop-2-yn-1-ol 97%
Supplier: Apollo Scientific
|
Warning
|
OR7333-1G
OR7333-250MG
OR7333-5G
APOSOR7333-1GEA
84.1
EUR
APOSOR7333-1G
APOSOR7333-250MG
APOSOR7333-5G
3-(2-Bromophenyl)prop-2-yn-1-ol 97%
3-(2-Bromophenyl)prop-2-yn-1-ol
Formula:
C₉H₇BrO MW: 211,055 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD04039054 CAS Number: 116509-98-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...