3-(3-Bromophenyl)prop-2-yn-1-ol 97%
Supplier: Apollo Scientific
|
Warning
|
OR7334-1G
OR7334-250MG
OR7334-5G
APOSOR7334-1GEA
212
EUR
APOSOR7334-1G
APOSOR7334-250MG
APOSOR7334-5G
3-(3-Bromophenyl)prop-2-yn-1-ol 97%
3-(3-Bromophenyl)prop-2-yn-1-ol
Formula:
C₉H₇BrO MW: 211,055 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD04039055 CAS Number: 170859-80-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...