3-(2-Chlorophenyl)prop-2-yn-1-ol
Supplier: Apollo Scientific
|
Warning
|
OR7380-1G
OR7380-250MG
OR7380-5G
APOSOR7380-1GEA
306
EUR
APOSOR7380-1G
APOSOR7380-250MG
APOSOR7380-5G
3-(2-Chlorophenyl)prop-2-yn-1-ol
3-(2-Chlorophenyl)prop-2-yn-1-ol
Formula:
C₉H₇ClO MW: 166.604 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04039063 CAS Number: 80151-26-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...