2-Chloro-5-(4-chlorophenyl)pyrimidine 95+%

Supplier: Apollo Scientific
Warning

OR7907-1G OR7907-5G OR7907-10G
APOSOR7907-1GEA 348 EUR
APOSOR7907-1G APOSOR7907-5G APOSOR7907-10G
2-Chloro-5-(4-chlorophenyl)pyrimidine 95+%
2-Chloro-5-(4-chlorophenyl)pyrimidine
Formula: C₁₀H₆Cl₂N₂
MW: 225,074 g/mol
Melting Pt: 215…216 °C
Storage Temperature: Ambient
MDL Number: MFCD03426061
CAS Number: 27956-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR