6-Chloro-2H-1-benzopyran-3-carbonyl chloride 95+%
Supplier: Apollo Scientific
OR8052-1G
OR8052-5G
APOSOR8052-1GEA
137
EUR
APOSOR8052-1G
APOSOR8052-5G
6-Chloro-2H-1-benzopyran-3-carbonyl chloride 95+%
6-Chloro-2H-1-benzopyran-3-carbonyl chloride
Formula:
C₁₀H₆Cl₂O₂ MW: 229,059 g/mol Melting Pt: 78…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD02091502 CAS Number: 306935-54-6 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...