1-Benzyl-4-methyl-1H-1,2,3-triazole

Supplier: Apollo Scientific
Warning

Synonyms: α-(4-Methyl-1H-1,2,3-triazol-1-yl)toluene, [(4-Methyl-1H-1,2,3-triazol-1-yl)methyl]benzene

OR8214-1G OR8214-250MG
APOSOR8214-1GEA 364 EUR
APOSOR8214-1G APOSOR8214-250MG
1-Benzyl-4-methyl-1H-1,2,3-triazole
1-Benzyl-4-methyl-1H-1,2,3-triazole
Formula: C₁₀H₁₁N₃
MW: 173,214 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08062394
CAS Number: 91258-00-3
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR