4-Aminotetrahydro-2H-thiopyran 95%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Aminothiane, Tetrahydro-2H-thiopyran-4-amine

OR8412-1G OR8412-5G OR8412-250MG
APOSOR8412-1GEA 72.5 EUR
APOSOR8412-1G APOSOR8412-5G APOSOR8412-250MG
4-Aminotetrahydro-2H-thiopyran 95%
4-Aminotetrahydro-2H-thiopyran
Formula: C₅H₁₁NS
MW: 117,213 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07371524
CAS Number: 21926-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR