2,4,6-Trimethylanisole
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Methoxymesitylene, 2-Methoxy-1,3,5-trimethylbenzene, Methyl 2,4,6-trimethylphenyl ether
OR8534-1G
OR8534-25G
OR8534-5G
APOSOR8534-1GEA
21.9
EUR
APOSOR8534-1G
APOSOR8534-25G
APOSOR8534-5G
2,4,6-Trimethylanisole
2,4,6-Trimethylanisole
Formula:
C₁₀H₁₄O MW: 150,218 g/mol Boiling Pt: 36…44 °C (0,5 mm Hg) Density: 0,953 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD03092887 CAS Number: 4028-66-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...