2-Bromo-5-methoxybenzhydrazide

Supplier: Apollo Scientific
Warning

Synonyms: 2-Bromo-5-methoxybenzohydrazide, 2-Bromo-5-methoxybenzoic acid hydrazide

OR8538-1G OR8538-10G
APOSOR8538-1GEA 29 EUR
APOSOR8538-1G APOSOR8538-10G
2-Bromo-5-methoxybenzhydrazide
2-Bromo-5-methoxybenzhydrazide
Formula: C₈H₉BrN₂O₂
MW: 245,073 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00117528
CAS Number: 112584-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR