N-Methyl-5-(aminomethyl)-7-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine 90%

Supplier: Apollo Scientific

OR8635-1G OR8635-250MG
APOSOR8635-1GEA 257 EUR
APOSOR8635-1G APOSOR8635-250MG
N-Methyl-5-(aminomethyl)-7-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine 90%
N-Methyl-5-(aminomethyl)-7-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine
Formula: C₈H₁₀BrNO₂S
MW: 264,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08690308
CAS Number: 886851-54-3
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR