2-(Pyrrolidin-1-yl)isonicotinaldehyde 97%

Supplier: Apollo Scientific
Danger

OR8674-1G OR8674-250MG
APOSOR8674-1GEA 364 EUR
APOSOR8674-1G APOSOR8674-250MG
2-(Pyrrolidin-1-yl)isonicotinaldehyde 97%
2-(Pyrrolidin-1-yl)isonicotinaldehyde
Formula: C₁₀H₁₂N₂O
MW: 176,215 g/mol
Melting Pt: 68…70 °C
Storage Temperature: Ambient
MDL Number: MFCD08690286
CAS Number: 898289-23-1
UN: 2928
ADR: 6.1+8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR