3-Bromo-2-methylbenzoyl chloride, tech

Supplier: Apollo Scientific
Danger

Synonyms: 2-Bromo-6-(chlorocarbonyl)toluene, 2-Bromo-6-(chloroformyl)toluene

OR8865-1G OR8865-5G OR8865-25G
APOSOR8865-1GEA 43.7 EUR
APOSOR8865-1G APOSOR8865-5G APOSOR8865-25G
3-Bromo-2-methylbenzoyl chloride, tech
3-Bromo-2-methylbenzoyl chloride
Formula: C₈H₆BrClO
MW: 233,49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08543466
CAS Number: 21900-48-1
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR