4-Methylsulphonylanisole 98%
Supplier: Apollo Scientific
Synonyms:
1-Methoxy-4-(methylsulphonyl)benzene
OR900068-1G
OR900068-5G
APOSOR900068-1GEA
56.7
EUR
APOSOR900068-1G
APOSOR900068-5G
4-Methylsulphonylanisole 98%
4-Methylsulphonylanisole
Formula:
C₈H₁₀O₃S MW: 186,228 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00025069 CAS Number: 3517-90-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...