Methyl (2E)-3,4,4-trimethylpent-2-enoate 98%
Supplier: Apollo Scientific
OR900482-1G
OR900482-5G
APOSOR900482-1GEA
270
EUR
APOSOR900482-1G
APOSOR900482-5G
Methyl (2E)-3,4,4-trimethylpent-2-enoate 98%
Methyl (2E)-3,4,4-trimethylpent-2-enoate
Formula:
C₉H₁₆O₂ MW: 156,222 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22586690 CAS Number: 16466-25-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...