4-Phenylcyclohex-3-en-1-amine 95%

Supplier: Apollo Scientific

OR900574-1G OR900574-250MG
APOSOR900574-1GEA 319 EUR
APOSOR900574-1G APOSOR900574-250MG
4-Phenylcyclohex-3-en-1-amine 95%
4-Phenylcyclohex-3-en-1-amine
Formula: C₁₂H₁₅N
MW: 173,254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01735224
CAS Number: 51171-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR