3-[(Pyrrolidin-1-yl)carbonyl]phenol 95%

Supplier: Apollo Scientific

OR900649-1G OR900649-25G OR900649-5G
APOSOR900649-1GEA 147 EUR
APOSOR900649-1G APOSOR900649-25G APOSOR900649-5G
3-[(Pyrrolidin-1-yl)carbonyl]phenol 95%
3-[(Pyrrolidin-1-yl)carbonyl]phenol
Formula: C₁₁H₁₃NO₂
MW: 191,226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07633879
CAS Number: 80917-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR