6-Chloro-8-methylquinoline 96%
Supplier: Apollo Scientific
OR901870-10G
OR901870-1G
OR901870-5G
APOSOR901870-10GEA
502
EUR
APOSOR901870-10G
APOSOR901870-1G
APOSOR901870-5G
6-Chloro-8-methylquinoline 96%
6-Chloro-8-methylquinoline
Formula:
C₁₀H₈ClN MW: 177,63 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10686879 CAS Number: 19655-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...