2-(Benzyloxy)pyridine 98%
Supplier: Apollo Scientific
OR902071-1G
OR902071-5G
APOSOR902071-1GEA
219
EUR
APOSOR902071-1G
APOSOR902071-5G
2-(Benzyloxy)pyridine 98%
2-(Benzyloxy)pyridine
Formula:
C₁₂H₁₁NO MW: 185,222 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09701434 CAS Number: 40864-08-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...