2,6-Dichlorophenyl isocyanide 95%

Supplier: Apollo Scientific

OR902397-1G OR902397-250MG
APOSOR902397-1GEA 807 EUR
APOSOR902397-1G APOSOR902397-250MG
2,6-Dichlorophenyl isocyanide 95%
2,6-Dichlorophenyl isocyanide
Formula: C₇H₃Cl₂N
MW: 172,011 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02664599
CAS Number: 6697-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR