Methyl 6-hydroxy-1H-indole-2-carboxylate 95%

Supplier: Apollo Scientific

OR902506-1G OR902506-250MG
APOSOR902506-1GEA 447 EUR
APOSOR902506-1G APOSOR902506-250MG
Methyl 6-hydroxy-1H-indole-2-carboxylate 95%
Methyl 6-hydroxy-1H-indole-2-carboxylate
Formula: C₁₀H₉NO₃
MW: 191,183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11053690
CAS Number: 116350-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR