4-(2-Chlorophenyl)-1,3-thiazole 95%

Supplier: Apollo Scientific

OR903036-1G OR903036-250MG
APOSOR903036-1GEA 865 EUR
APOSOR903036-1G APOSOR903036-250MG
4-(2-Chlorophenyl)-1,3-thiazole 95%
4-(2-Chlorophenyl)-1,3-thiazole
Formula: C₉H₆ClNS
MW: 195,669 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02665935
CAS Number: 383145-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR