2-(5-Methyl-2-furyl)piperidine 95%
Supplier: Apollo Scientific
OR903064-1G
OR903064-250MG
APOSOR903064-1GEA
865
EUR
APOSOR903064-1G
APOSOR903064-250MG
2-(5-Methyl-2-furyl)piperidine 95%
2-(5-Methyl-2-furyl)piperidine
Formula:
C₁₀H₁₅NO MW: 165,232 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07357570 CAS Number: 97073-18-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...