4-Phenyl-1,3-thiazole 96%

Supplier: Apollo Scientific

Synonyms: 4-Phenylthiazole

OR903354-1G OR903354-250MG
APOSOR903354-1GEA 78.3 EUR
APOSOR903354-1G APOSOR903354-250MG
4-Phenyl-1,3-thiazole 96%
4-Phenyl-1,3-thiazole
Formula: C₉H₇NS
MW: 161,224 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00092820
CAS Number: 1826-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR