(2,4,5-Trimethylphenyl)acetic acid 95%

Supplier: Apollo Scientific

OR903615-1G OR903615-250MG
APOSOR903615-1GEA 684 EUR
APOSOR903615-1G APOSOR903615-250MG
(2,4,5-Trimethylphenyl)acetic acid 95%
(2,4,5-Trimethylphenyl)acetic acid
Formula: C₁₁H₁₄O₂
MW: 178,228 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02664689
CAS Number: 3167-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR