2-Methoxyphenylisocyanide 95%

Supplier: Apollo Scientific

OR904051-100MG OR904051-1G OR904051-250MG
APOSOR904051-100MGEA 168 EUR
APOSOR904051-100MG APOSOR904051-1G APOSOR904051-250MG
2-Methoxyphenylisocyanide 95%
2-Methoxyphenylisocyanide
Formula: C₈H₇NO
MW: 133.147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02664590
CAS Number: 20771-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR