2-[(4-Methylphenyl)thio]ethanamine 95%
Supplier: Apollo Scientific
OR904253-1G
OR904253-250MG
OR904253-5G
APOSOR904253-1GEA
290
EUR
APOSOR904253-1G
APOSOR904253-250MG
APOSOR904253-5G
2-[(4-Methylphenyl)thio]ethanamine 95%
2-[(4-Methylphenyl)thio]ethanamine
Formula:
C₉H₁₃NS MW: 167,271 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04055859 CAS Number: 42404-23-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...