(S)-Methyl 1-Tritylaziridine-2-carboxylate

Supplier: Apollo Scientific
Warning

Synonyms: (S)-Methyl 1-tritylaziridine-2-carboxylate, Methyl (2S)-1-tritylaziridine-2-carboxylate

OR904264-5G OR904264-1G
APOSOR904264-5GEA 59.6 EUR
APOSOR904264-5G APOSOR904264-1G
(S)-Methyl 1-Tritylaziridine-2-carboxylate
(S)-Methyl 1-Tritylaziridine-2-carboxylate
Formula: C₂₃H₂₁NO₂
MW: 343.418 g/mol
Melting Pt: 124…128 °C
Storage Temperature: Ambient
MDL Number: MFCD01863736
CAS Number: 75154-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR