1-(6-Methyl-pyridin-2-yl)-1H-pyrrole-2-carbaldehyde 95%

Supplier: Apollo Scientific

OR904423-1G OR904423-250MG
APOSOR904423-1GEA 684 EUR
APOSOR904423-1G APOSOR904423-250MG
1-(6-Methyl-pyridin-2-yl)-1H-pyrrole-2-carbaldehyde 95%
1-(6-Methyl-pyridin-2-yl)-1H-pyrrole-2-carbaldehyde
Formula: C₁₁H₁₀N₂O
MW: 186,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02665161
CAS Number: 383136-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR