Methyl 6-hydroxypyrimidine-4-carboxylate 98%

Supplier: Apollo Scientific

Synonyms: Methyl 6-oxo-3,6-dihydropyrimidine-4-carboxylate

OR904554-1G OR904554-5G
APOSOR904554-1GEA 104 EUR
APOSOR904554-1G APOSOR904554-5G
Methyl 6-hydroxypyrimidine-4-carboxylate 98%
Methyl 6-hydroxypyrimidine-4-carboxylate
Formula: C₆H₆N₂O₃
MW: 154,123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00496785
CAS Number: 7399-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR