2-(5-Methyl-2-furyl)pyrrolidine 95%
Supplier: Apollo Scientific
OR904913-1G
OR904913-250MG
APOSOR904913-1GEA
806
EUR
APOSOR904913-1G
APOSOR904913-250MG
2-(5-Methyl-2-furyl)pyrrolidine 95%
2-(5-Methyl-2-furyl)pyrrolidine
Formula:
C₉H₁₃NO MW: 151,206 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06653530 CAS Number: 118248-27-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...