2-(1-Ethylpropyl)piperidine 95%

Supplier: Apollo Scientific

OR905802-1G OR905802-250MG
APOSOR905802-1GEA 328 EUR
APOSOR905802-1G APOSOR905802-250MG
2-(1-Ethylpropyl)piperidine 95%
2-(1-Ethylpropyl)piperidine
Formula: C₁₀H₂₁N
Storage Temperature: Ambient
MDL Number: MFCD02663646
CAS Number: 103906-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR