1-(Pyrrolidin-2-ylmethyl)piperidine 95%

Supplier: Apollo Scientific

OR905911-100MG OR905911-1G OR905911-250MG
APOSOR905911-100MGEA 190 EUR
APOSOR905911-100MG APOSOR905911-1G APOSOR905911-250MG
1-(Pyrrolidin-2-ylmethyl)piperidine 95%
1-(Pyrrolidin-2-ylmethyl)piperidine
Formula: C₁₀H₂₀N₂
MW: 168,279 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02663442
CAS Number: 112906-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR