2-(3-Chlorophenyl)piperidine 95%

Supplier: Apollo Scientific

OR906354-1G OR906354-250MG
APOSOR906354-1GEA 462 EUR
APOSOR906354-1G APOSOR906354-250MG
2-(3-Chlorophenyl)piperidine 95%
2-(3-Chlorophenyl)piperidine
Formula: C₁₁H₁₄ClN
MW: 195,689 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02663578
CAS Number: 383128-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR