2-Amino-4-cyclopropylthiophene-3-carbonitrile 98%
Supplier: Apollo Scientific
OR907319-10G
OR907319-1G
OR907319-5G
APOSOR907319-10GEA
754
EUR
APOSOR907319-10G
APOSOR907319-1G
APOSOR907319-5G
2-Amino-4-cyclopropylthiophene-3-carbonitrile 98%
2-Amino-4-cyclopropylthiophene-3-carbonitrile
Formula:
C₈H₈N₂S MW: 164,228 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08752930 CAS Number: 58124-28-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...