5-tert-butyl-2,3-dihydroinden-1-one 98%

Supplier: Apollo Scientific

Synonyms: 5-(tert-Butyl)-2,3-dihydro-1H-inden-1-one

OR907598-25G OR907598-5G
APOSOR907598-25GEA 437 EUR
APOSOR907598-25G APOSOR907598-5G
5-tert-butyl-2,3-dihydroinden-1-one 98%
5-tert-butyl-2,3-dihydroinden-1-one
Formula: C₁₃H₁₆O
MW: 188,266 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09033195
CAS Number: 4600-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR