methyl 4-cyano-3-methylbenzoate 95%

Supplier: Apollo Scientific

Synonyms: Methyl 4-cyano-3-methylbenzoate

OR907902-1G OR907902-5G
APOSOR907902-1GEA 77.1 EUR
APOSOR907902-1G APOSOR907902-5G
methyl 4-cyano-3-methylbenzoate 95%
methyl 4-cyano-3-methylbenzoate
Formula: C₁₀H₉NO₂
MW: 175,184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11655969
CAS Number: 25978-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR