1-(4-Chlorophenyl)cyclopropane-1-carboxamide 98%
Supplier: Apollo Scientific
OR908122-1G
OR908122-5G
APOSOR908122-1GEA
190
EUR
APOSOR908122-1G
APOSOR908122-5G
1-(4-Chlorophenyl)cyclopropane-1-carboxamide 98%
1-(4-Chlorophenyl)cyclopropane-1-carboxamide
Formula:
C₁₀H₁₀ClNO MW: 195,646 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10671794 CAS Number: 133284-47-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...