Ethyl 2-(2-oxo-1,3-oxazolidin-3-yl)acetate 95%

Supplier: Apollo Scientific

OR908187-1G OR908187-5G
APOSOR908187-1GEA 219 EUR
APOSOR908187-1G APOSOR908187-5G
Ethyl 2-(2-oxo-1,3-oxazolidin-3-yl)acetate 95%
Ethyl 2-(2-oxo-1,3-oxazolidin-3-yl)acetate
Formula: C₇H₁₁NO₄
MW: 173,167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11189498
CAS Number: 75125-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR