4-Bromo-3-(hydroxymethyl)benzoic acid 97%

Supplier: Apollo Scientific
Warning

OR908216-1G OR908216-250MG
APOSOR908216-1GEA 293 EUR
APOSOR908216-1G APOSOR908216-250MG
4-Bromo-3-(hydroxymethyl)benzoic acid 97%
4-Bromo-3-(hydroxymethyl)benzoic acid
Formula: C₈H₇BrO₃
Storage Temperature: Ambient
MDL Number: MFCD16249537
CAS Number: 790230-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR