2-(Cyclohexylethyl)boronic acid 95%

Supplier: Apollo Scientific

OR908303-1G OR908303-250MG
APOSOR908303-1GEA 335 EUR
APOSOR908303-1G APOSOR908303-250MG
2-(Cyclohexylethyl)boronic acid 95%
2-(Cyclohexylethyl)boronic acid
Formula: C₈H₁₇BO₂
MW: 156.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06212462
CAS Number: 105869-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR