5-Hydroxypridine-3-boronic acid 97%

Supplier: Apollo Scientific
Warning

OR908409-100MG OR908409-1G OR908409-250MG OR908409-2.5G OR908409-5G
APOSOR908409-100MGEA 26.1 EUR
APOSOR908409-100MG APOSOR908409-1G APOSOR908409-250MG APOSOR908409-2.5G APOSOR908409-5G
5-Hydroxypridine-3-boronic acid 97%
5-Hydroxypridine-3-boronic acid
Formula: C₅H₆BNO₃
MW: 138.917 g/mol
Melting Pt: 240…250 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13189132
CAS Number: 1208308-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR