4-chloro-2-ethoxyaniline 95%

Supplier: Apollo Scientific

OR908487-10G OR908487-1G OR908487-5G
APOSOR908487-10GEA 864 EUR
APOSOR908487-10G APOSOR908487-1G APOSOR908487-5G
4-chloro-2-ethoxyaniline 95%
4-chloro-2-ethoxyaniline
Formula: C₈H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD16732082
CAS Number: 846039-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR