1-(Cyclopropylmethyl)pyrazole-4-boronic acid 98%

Supplier: Apollo Scientific

OR908492-1G OR908492-250MG
APOSOR908492-1GEA 754 EUR
APOSOR908492-1G APOSOR908492-250MG
1-(Cyclopropylmethyl)pyrazole-4-boronic acid 98%
1-(Cyclopropylmethyl)pyrazole-4-boronic acid
Formula: C₇H₁₁BN₂O₂
MW: 165,985 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21332947
CAS Number: 1983202-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR