B-(3-Ethynylphenyl)-boronic acid 96%
Supplier: Apollo Scientific
Synonyms:
(3-Ethynylphenyl)boronic acid
OR908494-1G
OR908494-250MG
OR908494-5G
APOSOR908494-1GEA
270
EUR
APOSOR908494-1G
APOSOR908494-250MG
APOSOR908494-5G
B-(3-Ethynylphenyl)-boronic acid 96%
B-(3-Ethynylphenyl)-boronic acid
Formula:
C₈H₇BO₂ MW: 145,951 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17677372 CAS Number: 1189127-05-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...