(3-Methoxynaphthalen-1-yl)boronic acid ≥98%

Supplier: Apollo Scientific

OR908556-1G OR908556-250MG
APOSOR908556-1GEA 178 EUR
APOSOR908556-1G APOSOR908556-250MG
(3-Methoxynaphthalen-1-yl)boronic acid ≥98%
(3-Methoxynaphthalen-1-yl)boronic acid
Formula: C₁₁H₁₁BO₃
MW: 202,014 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18412862
CAS Number: 219834-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR