2-(Chloromethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%

Supplier: Apollo Scientific

OR908897-1G OR908897-5G
APOSOR908897-1GEA 59.6 EUR
APOSOR908897-1G APOSOR908897-5G
2-(Chloromethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%
2-(Chloromethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₇H₁₄BClO₂
MW: 176,449 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12405514
CAS Number: 83622-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR